missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyldimethylsilyloxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193802
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37391G Supplier TCI America Supplier No. B37391G
Only null left

Specifications

Chemical Name or Material 4-(tert-Butyldimethylsilyloxy)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 159191-56-7
Melting Point 207°C
Color White-Yellow
Molecular Formula C12H21BO3Si
Quantity 1 g
MDL Number MFCD03093888
Synonym 4-(tert-Butyldimethylsilyloxy)benzeneboronic Acid
InChI Key NVHHEADQQACSCJ-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)B(O)O
IUPAC Name {4-[(tert-butyldimethylsilyl)oxy]phenyl}boronic acid
Molecular Weight (g/mol) 252.19
PubChem CID 11107815
Formula Weight 252.19
Physical Form Crystalline Powder
Show More Show Less