missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyldimethylsilyloxy)cyclohexanone, 97%, Thermo Scientific Chemicals

Catalog No. AC426850050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426850050 Supplier Thermo Scientific Chemicals Supplier No. 426850050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55145-45-4 | Molecular Formula: C12H24O2Si | Molecular Weight: 228.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55145-45-4
Molecular Formula C12H24O2Si
Molecular Weight (g/mol) 228.41
MDL Number MFCD06411307
InChI Key HXKBGMNGSYGPRB-UHFFFAOYSA-N
Synonym 4-tert-butyldimethylsilyloxy cyclohexanone, 4-tert-butyldimethylsilyl oxy cyclohexanone, 4-tert-butyl dimethyl silyl oxy cyclohexanone, 4-tert-butyl-dimethyl-silanyloxy-cyclohexanone, 4-tert-butyldimethylsiloxy cyclohexanone, 4-tert-butyldimethylsilyloxy cyclohexanone meso form, 4-1,1-dimethylethyl dimethylsilyl oxy cyclohexanone, cyclohexanone, 4-1,1-dimethylethyl dimethylsilyl oxy, 4-tert-butyl dimethyl silyl oxycyclohexanone
PubChem CID 10609430
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxycyclohexan-1-one
SMILES CC(C)(C)[Si](C)(C)OC1CCC(=O)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9090g/mL
Boiling Point 85°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (O=)C6H9OSi(CH3)2C(CH3)3
Quantity 5 g
Specific Gravity 0.909
Formula Weight 228.41
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyldimethylsilyloxy)cyclohexanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only