accessibility menu, dialog, popup

UserName

H3CH3CCH3SiH3CCH3OO

CAS RN 55145-45-4

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]cyclohexan-1-one
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]cyclohexan-1-one
Molecular Weight (g/mol): 228.41
Molecular Formula: C12H24O2Si
InChi Key: HXKBGMNGSYGPRB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1CCC(=O)CC1
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)