missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyldimethylsilyloxy)-1-butyne 97.0+%, TCI America™

Catalog No. B50421ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50421ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B50421ML Supplier TCI America Supplier No. B50421ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78592-82-2
Molecular Formula C10H20OSi
Molecular Weight (g/mol) 184.354
MDL Number MFCD03427046
InChI Key NUZBJLXXTAOBPH-UHFFFAOYSA-N
Synonym (But-3-yn-1-yloxy)(tert-butyl)dimethylsilane
PubChem CID 4304567
IUPAC Name tert-butyl-but-3-ynoxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCC#C

Specifications

Color Yellow
Boiling Point 83°C
Quantity 1 mL
UN Number 1993
Formula Weight 184.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyldimethylsilyloxy)-1-butyne