CAS RN 78592-82-2
IUPAC Name:
(but-3-yn-1-yloxy)(tert-butyl)dimethylsilane
Synonyms:
(but-3-yn-1-yloxy)(tert-butyl)dimethylsilane
Molecular Weight (g/mol):
184.35
Molecular Formula:
C10H20OSi
InChi Key:
NUZBJLXXTAOBPH-UHFFFAOYSA-N
SMILES:
CC(C)(C)[Si](C)(C)OCCC#C
1
–
1
of
1
results
1
4-(tert-Butyldimethylsilyloxy)-1-butyne 97.0+%, TCI America™
CAS: 78592-82-2 Molecular Formula: C10H20OSi Molecular Weight (g/mol): 184.354 MDL Number: MFCD03427046 InChI Key: NUZBJLXXTAOBPH-UHFFFAOYSA-N Synonym: (But-3-yn-1-yloxy)(tert-butyl)dimethylsilane PubChem CID: 4304567 IUPAC Name: tert-butyl-but-3-ynoxy-dimethylsilane SMILES: CC(C)(C)[Si](C)(C)OCCC#C