missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexyl Acetate (cis- and trans- mixture) 96.0+%, TCI America™

Catalog No. p-7117028
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A072325ML 25 mL
A0723500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A072325ML Supplier TCI America Supplier No. A072325ML
Only null left

Chemical Identifiers

CAS 32210-23-4
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00019354
InChI Key MBZRJSQZCBXRGK-UHFFFAOYSA-N
Synonym Acetic Acid 4-tert-Butylcyclohexyl Ester
PubChem CID 36081
IUPAC Name 4-tert-butylcyclohexyl acetate
SMILES CC(=O)OC1CCC(CC1)C(C)(C)C

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylcyclohexyl Acetate (cis- and trans- mixture)
EINECSNumber (3)-2356
RTECSNumber AF7117000
TSCA Yes