accessibility menu, dialog, popup

UserName

CH3OOCH3H3CCH3

CAS RN 32210-23-4

IUPAC Name: 4-tert-butylcyclohexyl acetate
Synonyms: 4-tert-butylcyclohexyl acetate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: MBZRJSQZCBXRGK-UHFFFAOYSA-N
SMILES: CC(=O)OC1CCC(CC1)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results