missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanone 97.0+%, TCI America™

Catalog No. p-7118834
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B107425G 25 g
B1074100G 100 g
B1074500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B107425G Supplier TCI America Supplier No. B107425G
Only null left

Chemical Identifiers

CAS 98-53-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001642
InChI Key YKFKEYKJGVSEIX-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
PubChem CID 7392
IUPAC Name 4-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCC(=O)CC1

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 116°C
Quantity 25 g
Formula Weight 154.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcyclohexanone
RTECSNumber GW1140000
TSCA Yes