missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanone, 99%

Catalog No. AAA1561222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561222 100 g
AAA1561236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1561222 Supplier Thermo Scientific Chemicals Supplier No. A1561222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-53-3 | C10H18O | 154.253 g/mol

4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.

Solubility
Soluble in alcohol, ethanol (0.5g/10 mL). Insoluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 98-53-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001642
InChI Key YKFKEYKJGVSEIX-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
PubChem CID 7392
IUPAC Name 4-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCC(=O)CC1

Specifications

Melting Point 47°C to 51°C
Density 0.893
Boiling Point 113°C to 116°C (20 mmHg)
Flash Point 96°C (204°F)
Quantity 100 g
Beilstein 507309
Solubility Information Soluble in alcohol,ethanol (0.5g/10 mL). Insoluble in water.
Formula Weight 154.25
Percent Purity 99%
Chemical Name or Material 4-tert-Butylcyclohexanone
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-678-5
RTECSNumber GW1140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only