missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%

CAS: 5451-55-8 | C11H20O2 | 184.279 g/mol

$109.16 - $332.52

Chemical Identifiers

CAS 5451-55-8
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00042622
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
Synonym trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL0824806
View Documents Promotion Details
N/A 5 g
Each for $109.16
Only null left
 
AAL0824814
View Documents Promotion Details
N/A 25 g
Each for $332.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5451-55-8
184.279
QVQKEGYITJBHRQ-UHFFFAOYSA-N
136759
CC(C)(C)C1CCC(CC1)C(=O)O
C11H20O2
MFCD00042622
trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
4-tert-butylcyclohexane-1-carboxylic acid

Specifications

5451-55-8
MFCD00042622
trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n
CC(C)(C)C1CCC(CC1)C(=O)O
184.279
184.28
4-tert-Butylcyclohexanecarboxylic acid, predominantly trans
C11H20O2
2043187
QVQKEGYITJBHRQ-UHFFFAOYSA-N
4-tert-butylcyclohexane-1-carboxylic acid
136759
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only