accessibility menu, dialog, popup

UserName

CAS RN 5451-55-8

IUPAC Name: 4-tert-butylcyclohexane-1-carboxylic acid
Synonyms: 4-tert-butylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: QVQKEGYITJBHRQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)