missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America B128525G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C11H20O2 | |
| MFCD00042622 | |
| trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n | |
| 4-tert-butylcyclohexane-1-carboxylic acid |
Specifications
| 4-tert-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) | |
| White | |
| 25 g | |
| trans-4-tert-butylcyclohexanecarboxylic acid, 4-tert-butylcyclohexanecarboxylic acid, cis-4-tert-butylcyclohexanecarboxylic acid, trans-4-tert-butyl cyclohexanecarboxylic acid, 4-tert-butyl cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, cis, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl, cis-4-tert-butylcyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-1,1-dimethylethyl-, trans, qvqkegyitjbhrq-dtorhvgosa-n | |
| CC(C)(C)C1CCC(CC1)C(=O)O | |
| 184.279 | |
| 184.28 | |
| Crystalline Powder |
| 5451-55-8 | |
| C11H20O2 | |
| MFCD00042622 | |
| QVQKEGYITJBHRQ-UHFFFAOYSA-N | |
| 4-tert-butylcyclohexane-1-carboxylic acid | |
| 136759 | |
| ≥98.0% (GC,T) |
Safety and Handling
EINECSNumber : (3)-3723
TSCA : No