missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[6]arene, 96%

Catalog No. AAL1447303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1447303 1 g
AAL1447306 5 g
AAL1447314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1447303 Supplier Thermo Scientific Chemicals Supplier No. L1447303

CAS: 78092-53-2 | C66H84O6 | 973.392 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78092-53-2
Molecular Formula C66H84O6
Molecular Weight (g/mol) 973.392
MDL Number MFCD00075464
InChI Key UOEYZAXKBKAKRO-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 6 arene, unii-b9og173dmy, b9og173dmy, 5,11,17,23,29,35-hexa-tert-butylcalix 6 arene-37,38,39,40,41,42-hexol, hexa-tert-butyl hexahydroxy calix 6 arene, calx-b 6, p-t-butylcalix 6 arene, tert-butylcalix 6 arene, p-tert-butylcalix-6-arene
PubChem CID 335356
ChEBI CHEBI:51251
SMILES CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=C(C(=CC(=C6)C(C)(C)C)CC7=C(C(=CC(=C7)C(C)(C)C)C2)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)O

Specifications

Melting Point 377°C to 381°C
Beilstein 2611918
Quantity 1 g
Formula Weight 973.4
Percent Purity 96%
Chemical Name or Material 4-tert-Butylcalix[6]arene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only