missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzylamine 98.0+%, TCI America™
Supplier: TCI America B158725ML
Specifications
| 4-tert-Butylbenzylamine | |
| Colorless | |
| C11H17N | |
| 25 mL | |
| 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine | |
| CC(C)(C)C1=CC=C(C=C1)CN | |
| 163.264 | |
| 163.26 | |
| Liquid |
| 39895-55-1 | |
| 124°C | |
| MFCD00040754 | |
| 2735 | |
| MPWSRGAWRAYBJK-UHFFFAOYSA-N | |
| (4-tert-butylphenyl)methanamine | |
| 2735655 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 39895-55-1 | |
| 163.264 | |
| MPWSRGAWRAYBJK-UHFFFAOYSA-N | |
| 2735655 | |
| CC(C)(C)C1=CC=C(C=C1)CN |
| C11H17N | |
| MFCD00040754 | |
| 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine | |
| (4-tert-butylphenyl)methanamine |
Safety and Handling
EINECSNumber : (3)-4120
TSCA : No