missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzylamine 98.0+%, TCI America™

Catalog No. B15875ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15875ML 5 mL
B158725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15875ML Supplier TCI America Supplier No. B15875ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39895-55-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00040754
InChI Key MPWSRGAWRAYBJK-UHFFFAOYSA-N
Synonym 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine
PubChem CID 2735655
IUPAC Name (4-tert-butylphenyl)methanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CN

Specifications

Color Colorless
Boiling Point 124°C
Quantity 5 mL
UN Number 2735
Formula Weight 163.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzylamine
EINECSNumber (3)-4120
TSCA No