missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzylamine, 98%
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible or difficult to mix with water.
Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible or difficult to mix with water.
Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Density | 0.927 |
| Boiling Point | 124°C (16 mmHg) |
| Flash Point | 107°C (224°F) |
| Refractive Index | 1.521 |
| Quantity | 5 g |
| UN Number | UN2735 |
| Beilstein | 2961797 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 163.26 |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 254-681-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only