Learn More
4-tert-Butylbenzylamine, 97%
CAS: 39895-55-1 | C11H17N | 163.26 g/mol
$274.72 - $274.72
Chemical Identifiers
| CAS | 39895-55-1 |
|---|---|
| Molecular Formula | C11H17N |
| Molecular Weight (g/mol) | 163.26 |
| MDL Number | MFCD00040754 |
| InChI Key | MPWSRGAWRAYBJK-UHFFFAOYSA-N |
| Synonym | 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine |
| PubChem CID | 2735655 |
| IUPAC Name | (4-tert-butylphenyl)methanamine |
| SMILES | CC(C)(C)C1=CC=C(C=C1)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC391010050
|
Thermo Scientific Chemicals
391010050 |
5 g | Glass bottle |
Each for $274.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39895-55-1 | |
| 163.26 | |
| MPWSRGAWRAYBJK-UHFFFAOYSA-N | |
| 2735655 | |
| CC(C)(C)C1=CC=C(C=C1)CN |
| C11H17N | |
| MFCD00040754 | |
| 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine | |
| (4-tert-butylphenyl)methanamine |
Specifications
| 39895-55-1 | |
| 100.0 | |
| 0.9270g/mL | |
| 107°C | |
| 96% min. (GC) | |
| C11H17N | |
| NH2CH2C6H4C(CH3) | |
| 5 g | |
| 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine | |
| CC(C)(C)C1=CC=C(C=C1)CN | |
| 163.26 | |
| 163.26 | |
| Liquid |
| 96.0 | |
| Yellow | |
| 235°C to 236°C | |
| Authentic | |
| Glass bottle | |
| 1.5200 to 1.522 | |
| MFCD00040754 | |
| 0.927 | |
| MPWSRGAWRAYBJK-UHFFFAOYSA-N | |
| (4-tert-butylphenyl)methanamine | |
| 2735655 | |
| 97% | |
| 4-tert-Butylbenzylamine |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 254-681-6
RUO – Research Use Only