missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl bromide, 97%

Catalog No. AAH2699106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2699106 5 g
AAH2699114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2699106 Supplier Thermo Scientific Chemicals Supplier No. H2699106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18880-00-7 | C11H15Br | 227.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18880-00-7
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00000180
InChI Key QZNQSIHCDAGZIA-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide
PubChem CID 87836
IUPAC Name 1-(bromomethyl)-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)CBr

Specifications

Melting Point 8°C to 12°C
Density 1.236
Boiling Point 93°C to 94°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5465
Quantity 5 g
UN Number UN3265
Beilstein 471674
Formula Weight 227.14
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 242-643-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only