missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzophenone, 98%

Catalog No. AAL0638606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0638606 5 g
AAL0638603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0638606 Supplier Thermo Scientific Chemicals Supplier No. L0638606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22679-54-5 | C17H18O | 238.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22679-54-5
Molecular Formula C17H18O
Molecular Weight (g/mol) 238.33
MDL Number MFCD00017255
InChI Key DFYJCXSOGSYMAJ-UHFFFAOYSA-N
Synonym 4-tert-butylbenzophenone, 4-t-butyl benzophenone, 4-tert-butyl phenyl phenyl methanone, 4-tert-butyl-benzophenone, 4-tert-butylphenyl-phenylmethanone, 4-tert-butylphenyl phenyl methanone, methanone, 4-1,1-dimethylethyl phenyl phenyl, acmc-20aman, 4-tert.-butylbenzophenone
PubChem CID 89790
IUPAC Name (4-tert-butylphenyl)-phenylmethanone
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 38°C to 42°C
Boiling Point 168°C to 170°C (2 mmHg)
Flash Point 150°C (302°F)
Quantity 5 g
Beilstein 2100075
Formula Weight 238.32
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzophenone
EINECSNumber 245-154-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only