missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoic anhydride, 95%

Catalog No. AAH3242206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3242206 5 g
AAH3242203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3242206 Supplier Thermo Scientific Chemicals Supplier No. H3242206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22201-45-2 | C22H26O3 | 338.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22201-45-2
Molecular Formula C22H26O3
Molecular Weight (g/mol) 338.45
MDL Number MFCD09757530
InChI Key KKDPRVYYZZUPLR-UHFFFAOYSA-N
Synonym 4-tert-butylbenzoic anhydride, 4-tert-butylbenzoyl 4-tert-butylbenzoate, bis 4-tert-butylbenzoic acid anhydride, benzoic acid,4-1,1-dimethylethyl-, anhydride with 4-1,1-dimethylethyl benzoic acid
PubChem CID 14962255
IUPAC Name (4-tert-butylbenzoyl) 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 75°C to 81°C
Quantity 5 g
Formula Weight 338.45
Percent Purity 95%
Chemical Name or Material 4-tert-Butylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only