missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoic Acid 99.0+%, TCI America™

Catalog No. B0717500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0717500G 500 g
B071725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0717500G Supplier TCI America Supplier No. B0717500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-73-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00002563
InChI Key KDVYCTOWXSLNNI-UHFFFAOYSA-N
Synonym 4-tert-butyl benzoic acid, tbba, p-tert-butylbenzoic acid, benzoic acid, 4-1,1-dimethylethyl, 4-t-butylbenzoic acid, benzoic acid, p-tert-butyl, 4-tert-butyl-benzoic acid, p-t-butylbenzoic acid, 4-tert-butylbenzoicacid, unii-43z7t3vn0r
PubChem CID 7403
ChEBI CHEBI:34443
IUPAC Name 4-tert-butylbenzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 167°C
Color White
Quantity 500 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DG4708000
TSCA Yes