Learn More
4-tert-Butylbenzoic acid, 99%
CAS: 98-73-7 | C11H14O2 | 178.23 g/mol
Supplier: Thermo Scientific Chemicals A139970B
Description
4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
| 4-tert-Butylbenzoic acid | |
| 164°C to 168°C | |
| 180°C (356°F) | |
| MFCD00002563 | |
| UN2811 | |
| 4-tert-butyl benzoic acid, tbba, p-tert-butylbenzoic acid, benzoic acid, 4-1,1-dimethylethyl, 4-t-butylbenzoic acid, benzoic acid, p-tert-butyl, 4-tert-butyl-benzoic acid, p-t-butylbenzoic acid, 4-tert-butylbenzoicacid, unii-43z7t3vn0r | |
| KDVYCTOWXSLNNI-UHFFFAOYSA-N | |
| 4-tert-butylbenzoic acid | |
| 7403 | |
| 178.23 |
| 98-73-7 | |
| 280°C (decomposition) | |
| C11H14O2 | |
| 1000 g | |
| 607545 | |
| Insoluble in water. | |
| CC(C)(C)C1=CC=C(C=C1)C(O)=O | |
| 178.23 | |
| CHEBI:34443 | |
| ≥99% |
Chemical Identifiers
| 98-73-7 | |
| 178.23 | |
| KDVYCTOWXSLNNI-UHFFFAOYSA-N | |
| 7403 | |
| 4-tert-butylbenzoic acid |
| C11H14O2 | |
| MFCD00002563 | |
| 4-tert-butyl benzoic acid, tbba, p-tert-butylbenzoic acid, benzoic acid, 4-1,1-dimethylethyl, 4-t-butylbenzoic acid, benzoic acid, p-tert-butyl, 4-tert-butyl-benzoic acid, p-t-butylbenzoic acid, 4-tert-butylbenzoicacid, unii-43z7t3vn0r | |
| CHEBI:34443 | |
| CC(C)(C)C1=CC=C(C=C1)C(O)=O |
Safety and Handling
GHS H Statement
H311-H360-H372-H373-H302
Toxic in contact with skin.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P361-P363-P501c
H302-H311-H332-H360-H372-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-696-3
RTECSNumber : DG4708000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only