missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylaniline 98.0+%, TCI America™

Catalog No. B19005ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19005ML 5 mL
B190025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19005ML Supplier TCI America Supplier No. B19005ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 769-92-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00007899
InChI Key WRDWWAVNELMWAM-UHFFFAOYSA-N
Synonym 4-tert-butyl aniline, p-tert-butylaniline, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, 4-t-butylaniline, unii-47h9440zow, 4-t-butylbenzeneamine, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine
PubChem CID 69861
IUPAC Name 4-tert-butylaniline
SMILES CC(C)(C)C1=CC=C(C=C1)N

Specifications

Boiling Point 230°C
Quantity 5 mL
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylaniline
EINECSNumber (3)-0206
TSCA No
Recommended Storage Refrigerator