missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylaniline, 98+%

Catalog No. AAA1092506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1092506 5 g
AAA1092514 25 g
AAA1092522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1092506 Supplier Thermo Scientific Chemicals Supplier No. A1092506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 769-92-6 | C10H15N | 149.237 g/mol

4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4?,4?-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4′,4″-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong acids, acid chlorides, acid anhydrides and chloroformates.

Chemical Identifiers

CAS 769-92-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00007899
InChI Key WRDWWAVNELMWAM-UHFFFAOYSA-N
Synonym 4-tert-butyl aniline, p-tert-butylaniline, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, 4-t-butylaniline, unii-47h9440zow, 4-t-butylbenzeneamine, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine
PubChem CID 69861
IUPAC Name 4-tert-butylaniline
SMILES CC(C)(C)C1=CC=C(C=C1)N

Specifications

Melting Point 15°C to 16°C
Density 0.941
Boiling Point 235°C
Flash Point 101°C (213°F)
Refractive Index 1.538
Quantity 5 g
Beilstein 2205786
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 149.23
Percent Purity 99%
Chemical Name or Material 4-tert-Butylaniline
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-215-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only