Learn More
4-(tert-Butyl)-o-phenylenediamine, 97%
CAS: 68176-57-8 | C10H16N2 | 164.252 g/mol
$103.27 - $361.89
Chemical Identifiers
| CAS | 68176-57-8 |
|---|---|
| Molecular Formula | C10H16N2 |
| Molecular Weight (g/mol) | 164.252 |
| MDL Number | MFCD00052695 |
| InChI Key | WLOSFXSXVXTKBU-UHFFFAOYSA-N |
| PubChem CID | 432708 |
| IUPAC Name | 4-tert-butylbenzene-1,2-diamine |
| SMILES | CC(C)(C)C1=CC(=C(C=C1)N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3220503
|
Thermo Scientific Chemicals
H3220503 |
1 g |
Each for $103.27
|
|
|||||
|
AAH3220506
|
Thermo Scientific Chemicals
H3220506 |
5 g |
Each for $361.89
|
|
|||||
Description
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.
Solubility
Soluble in most common organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 68176-57-8 | |
| 164.252 | |
| WLOSFXSXVXTKBU-UHFFFAOYSA-N | |
| 4-tert-butylbenzene-1,2-diamine |
| C10H16N2 | |
| MFCD00052695 | |
| 432708 | |
| CC(C)(C)C1=CC(=C(C=C1)N)N |
Specifications
| 68176-57-8 | |
| C10H16N2 | |
| 1 g | |
| Air Sensitive | |
| Soluble in most common organic solvents. | |
| CC(C)(C)C1=CC(=C(C=C1)N)N | |
| 164.252 | |
| 164.25 | |
| 4-(tert-Butyl)-o-phenylenediamine |
| 98°C to 100°C | |
| MFCD00052695 | |
| UN2811 | |
| 4-tert-butyl benzene-1,2-diamine, 4-tert-butyl-1,2-diaminobenzene, 1,2-diamino-4-tert-butylbenzene, 4-tert-butyl-o-phenylenediamine, 1,2-benzenediamine,4-1,1-dimethylethyl, 1,2-benzenediamine, 4-1,1-dimethylethyl, pubchem22919, 4-tert-butyl-diaminobenzene, 4-t-butylbenzene-1,2-diamine, 4-tert-butylbenzen-1,2-diamine | |
| WLOSFXSXVXTKBU-UHFFFAOYSA-N | |
| 4-tert-butylbenzene-1,2-diamine | |
| 432708 | |
| 97% |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
H302+H332-H311
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only