missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-o-phenylenediamine, 97%

Catalog No. AAH3220503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3220503 1 g
AAH3220506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3220503 Supplier Thermo Scientific Chemicals Supplier No. H3220503

CAS: 68176-57-8 | C10H16N2 | 164.252 g/mol

4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

Solubility
Soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 68176-57-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00052695
InChI Key WLOSFXSXVXTKBU-UHFFFAOYSA-N
PubChem CID 432708
IUPAC Name 4-tert-butylbenzene-1,2-diamine
SMILES CC(C)(C)C1=CC(=C(C=C1)N)N

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
UN Number UN2811
Sensitivity Air Sensitive
Synonym 4-tert-butyl benzene-1,2-diamine, 4-tert-butyl-1,2-diaminobenzene, 1,2-diamino-4-tert-butylbenzene, 4-tert-butyl-o-phenylenediamine, 1,2-benzenediamine,4-1,1-dimethylethyl, 1,2-benzenediamine, 4-1,1-dimethylethyl, pubchem22919, 4-tert-butyl-diaminobenzene, 4-t-butylbenzene-1,2-diamine, 4-tert-butylbenzen-1,2-diamine
Solubility Information Soluble in most common organic solvents.
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)-o-phenylenediamine
Hazard Category H302+H332-H311
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only