missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™

Catalog No. p-7123255
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39351G 1 g
B39355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39351G Supplier TCI America Supplier No. B39351G
Only null left

Chemical Identifiers

CAS 1676-90-0
Molecular Formula C13H23NO6
Molecular Weight (g/mol) 289.328
MDL Number MFCD00076912
InChI Key PHJDCONJXLIIPW-QMMMGPOBSA-N
Synonym boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh
PubChem CID 7010636
IUPAC Name (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Melting Point 63°C
Color White
Quantity 1 g
Formula Weight 289.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate
TSCA No
Recommended Storage Refrigerator