missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™
Supplier: TCI America B315025G
Specifications
| 4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate | |
| 150°C | |
| C23H25NO6 | |
| 25 g | |
| FODJWPHPWBKDON-IBGZPJMESA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
| 2724635 | |
| ≥98.0% (HPLC,T) |
| 71989-14-5 | |
| White-Yellow | |
| MFCD00037131 | |
| fmoc-asp otbu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-asp obut, fmoc-l-aspartic acid beta-tert-butyl ester, fmoc-asp ot-bu-oh, fmoc-l-asp tbu, fmocasp otbu-oh, fmoc-asp obu-oh, fmoc-asp tbu-oh, fmoc-l-aspartic acid 4-tert-butyl ester | |
| CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 411.454 | |
| 411.45 | |
| Crystalline Powder |
Chemical Identifiers
| 71989-14-5 | |
| 411.454 | |
| FODJWPHPWBKDON-IBGZPJMESA-N | |
| 2724635 | |
| CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C23H25NO6 | |
| MFCD00037131 | |
| fmoc-asp otbu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-asp obut, fmoc-l-aspartic acid beta-tert-butyl ester, fmoc-asp ot-bu-oh, fmoc-l-asp tbu, fmocasp otbu-oh, fmoc-asp obu-oh, fmoc-asp tbu-oh, fmoc-l-aspartic acid 4-tert-butyl ester | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
Safety and Handling
TSCA : No