accessibility menu, dialog, popup

UserName

CAS RN 71989-14-5

IUPAC Name: (2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoate
Synonyms: (2S)-4-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoate
Molecular Weight (g/mol): 410.45
Molecular Formula: C23H24NO6
InChi Key: FODJWPHPWBKDON-IBGZPJMESA-M
SMILES: CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)