missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™

Catalog No. B31505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31505G 5 g
B315025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31505G Supplier TCI America Supplier No. B31505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71989-14-5
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.454
MDL Number MFCD00037131
InChI Key FODJWPHPWBKDON-IBGZPJMESA-N
Synonym fmoc-asp otbu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-asp obut, fmoc-l-aspartic acid beta-tert-butyl ester, fmoc-asp ot-bu-oh, fmoc-l-asp tbu, fmocasp otbu-oh, fmoc-asp obu-oh, fmoc-asp tbu-oh, fmoc-l-aspartic acid 4-tert-butyl ester
PubChem CID 2724635
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 411.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate