Learn More
4-(tert-Butyl)benzylbromide, 97%
CAS: 18880-00-7 | C11H15Br | 227.14 g/mol
$67.05 - $236.29
Chemical Identifiers
| CAS | 18880-00-7 |
|---|---|
| Molecular Formula | C11H15Br |
| Molecular Weight (g/mol) | 227.14 |
| MDL Number | MFCD00000180 |
| InChI Key | QZNQSIHCDAGZIA-UHFFFAOYSA-N |
| Synonym | 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide |
| PubChem CID | 87836 |
| IUPAC Name | 1-(bromomethyl)-4-tert-butylbenzene |
| SMILES | CC(C)(C)C1=CC=C(C=C1)CBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC345500050
|
Thermo Scientific Chemicals
345500050 |
5 g | Glass bottle |
Each for $67.05
|
|
||||
|
AC345500250
|
Thermo Scientific Chemicals
345500250 |
25 g | Glass bottle |
Each for $236.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18880-00-7 | |
| 227.14 | |
| QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
| 87836 | |
| CC(C)(C)C1=CC=C(C=C1)CBr |
| C11H15Br | |
| MFCD00000180 | |
| 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
| 1-(bromomethyl)-4-tert-butylbenzene |
Specifications
| 18880-00-7 | |
| 100.0 | |
| Yellow | |
| 90°C to 94°C (1.5 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5445 to 1.5465 | |
| MFCD00000180 | |
| 1.236 | |
| QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-tert-butylbenzene | |
| 87836 | |
| 97% | |
| 4-(tert-Butyl)benzylbromide |
| 96.0 | |
| 8°C to 12°C | |
| 1.2360g/mL | |
| >109°C | |
| 96% min. (Argentometry) | |
| C11H15Br | |
| (CH3)3CC6H4CH2Br | |
| 5 g | |
| 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
| CC(C)(C)C1=CC=C(C=C1)CBr | |
| 227.14 | |
| 227.14 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 242-643-1
RUO – Research Use Only