missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)-2-chloropyridine 98.0+%, TCI America™
$281.48 - $961.86
Chemical Identifiers
| CAS | 81167-60-4 |
|---|---|
| Molecular Formula | C9H12ClN |
| Molecular Weight (g/mol) | 169.65 |
| MDL Number | MFCD12131129 |
| InChI Key | UGGNULHQVUWRGQ-UHFFFAOYSA-N |
| PubChem CID | 57345981 |
| IUPAC Name | 4-tert-butyl-2-chloropyridine |
| SMILES | CC(C)(C)C1=CC(Cl)=NC=C1 |
Chemical Identifiers
| 81167-60-4 | |
| 169.65 | |
| UGGNULHQVUWRGQ-UHFFFAOYSA-N | |
| 4-tert-butyl-2-chloropyridine |
| C9H12ClN | |
| MFCD12131129 | |
| 57345981 | |
| CC(C)(C)C1=CC(Cl)=NC=C1 |
Specifications
| 81167-60-4 | |
| MFCD12131129 | |
| UGGNULHQVUWRGQ-UHFFFAOYSA-N | |
| 4-tert-butyl-2-chloropyridine | |
| 57345981 | |
| ≥98.0% (GC) | |
| 4-(tert-Butyl)-2-chloropyridine |
| C9H12ClN | |
| 1 g | |
| CC(C)(C)C1=CC(Cl)=NC=C1 | |
| 169.65 | |
| 169.65 | |
| Liquid |
Safety and Handling
TSCA : No