missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)-2-chloropyridine 98.0+%, TCI America™
Supplier: TCI America B54661G
Specifications
| 4-(tert-Butyl)-2-chloropyridine | |
| C9H12ClN | |
| 1 g | |
| CC(C)(C)C1=CC(Cl)=NC=C1 | |
| 169.65 | |
| 169.65 | |
| Liquid |
| 81167-60-4 | |
| MFCD12131129 | |
| UGGNULHQVUWRGQ-UHFFFAOYSA-N | |
| 4-tert-butyl-2-chloropyridine | |
| 57345981 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 81167-60-4 | |
| 169.65 | |
| UGGNULHQVUWRGQ-UHFFFAOYSA-N | |
| 4-tert-butyl-2-chloropyridine |
| C9H12ClN | |
| MFCD12131129 | |
| 57345981 | |
| CC(C)(C)C1=CC(Cl)=NC=C1 |
Safety and Handling
TSCA : No