missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-2,6-dinitrophenol, 97%

Catalog No. AAL0867006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0867006 5 g
AAL0867014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0867006 Supplier Thermo Scientific Chemicals Supplier No. L0867006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4097-49-8 | C10H12N2O5 | 240.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4097-49-8
Molecular Formula C10H12N2O5
Molecular Weight (g/mol) 240.22
MDL Number MFCD00051969
InChI Key NJBDTWSOYUZQPM-UHFFFAOYSA-N
Synonym 4-tert-butyl-2,6-dinitrophenol, 2,6-dinitro-4-tert-butylphenol, unii-m5s512rl03, phenol, 4-1,1-dimethylethyl-2,6-dinitro, 2,6-dinitro-p-tert-butyl phenol, phenol, 4-tert-butyl-2,6-dinitro, acmc-1am7s, 2,6-dinitro-4-t-butylphenol, 4-t-butyl-2,6-dinitrophenol
PubChem CID 20042
IUPAC Name 4-tert-butyl-2,6-dinitrophenol
SMILES CC(C)(C)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 93°C to 95°C
Quantity 5 g
UN Number UN2811
Beilstein 1986466
Formula Weight 240.22
Percent Purity 97%
Chemical Name or Material 4-tert-Butyl-2,6-dinitrophenol
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 223-856-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only