missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-1-(3-sulfopropyl)pyridinium Hydroxide Inner Salt Hydrate 98.0+%, TCI America™

Catalog No. B40301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40301G Supplier TCI America Supplier No. B40301G
Only null left

Chemical Identifiers

CAS 570412-84-9
Molecular Formula C12H19NO3S
Molecular Weight (g/mol) 257.348
MDL Number MFCD06411435
InChI Key QMGYGZCGWOKPGR-UHFFFAOYSA-N
Synonym 3-(4-tert-Butyl-1-pyridinio)propanesulfonate, NDSB 256-4T
PubChem CID 16760575
IUPAC Name 3-(4-tert-butylpyridin-1-ium-1-yl)propane-1-sulfonate
SMILES CC(C)(C)C1=CC=[N+](C=C1)CCCS(=O)(=O)[O-]

Specifications

Color White
Quantity 1 g
Formula Weight 257.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl-1-(3-sulfopropyl)pyridinium Hydroxide Inner Salt Hydrate [for Biochemical Research]