accessibility menu, dialog, popup

UserName

CAS RN 570412-84-9

CH3H3CCH3NSOOO

CAS RN 570412-84-9

IUPAC Name: 4-tert-butyl-1-(3-sulfonatopropyl)pyridin-1-ium
Synonyms: 4-tert-butyl-1-(3-sulfonatopropyl)pyridin-1-ium
Molecular Weight (g/mol): 257.35
Molecular Formula: C12H19NO3S
InChi Key: QMGYGZCGWOKPGR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=[N+](CCCS([O-])(=O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results