missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. B32515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32515G Supplier TCI America Supplier No. B32515G

Chemical Identifiers

CAS 162046-58-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
InChI Key AZEKNJGFCSHZID-UHFFFAOYSA-N
Synonym boc-tranexamic acid, 4-tert-butoxycarbonyl amino methyl cyclohexanecarboxylic acid, boc-4-amc-oh, trans-4-tert-butoxycarbonylaminomethyl cyclohexanecarboxylic acid, 4-tert-butoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, trans-4-n-boc-aminomethyl-cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonyl amino methyl cyclohexanecarboxylic acid, 4-boc-aminomethyl cyclohexanecarboxylic acid, cis-4-boc-aminomethyl-cyclohexyl carboxylic acid
PubChem CID 736918
IUPAC Name 4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 257.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture)