accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNOHO

CAS RN 162046-58-4

IUPAC Name: 4-({[(tert-butoxy)carbonyl]amino}methyl)cyclohexane-1-carboxylic acid
Synonyms: 4-{[(tert-butoxycarbonyl)amino]methyl}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 257.33
Molecular Formula: C13H23NO4
InChi Key: AZEKNJGFCSHZID-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results