missing translation for 'onlineSavingsMsg'
Learn More

4-[(tert-Butoxycarbonylamino)methyl]benzoic Acid 98.0+%, TCI America™

Catalog No. B43055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43055G 5 g
B43051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43055G Supplier TCI America Supplier No. B43055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33233-67-9
Molecular Formula C13H16NO4
Molecular Weight (g/mol) 250.28
MDL Number MFCD00228182
InChI Key LNKHBRDWRIIROP-UHFFFAOYSA-M
Synonym 4-tert-butoxycarbonylamino methyl benzoic acid, 4-tert-butoxycarbonyl amino methyl benzoic acid, 4-tert-butoxy carbonyl amino methyl benzoic acid, boc-4-aminomethyl-benzoic acid, 4-boc-aminomethyl-benzoic acid, 4-tert-butoxycarbonylaminomethyl benzoic acid, 4-n-tert-butyloxycarbonyl amino methyl benzoic acid, benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl
PubChem CID 853605
IUPAC Name 4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]benzoic Acid