accessibility menu, dialog, popup

UserName

CAS RN 33233-67-9

IUPAC Name: 4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate
Synonyms: 4-{[(tert-butoxycarbonyl)amino]methyl}benzoate
Molecular Weight (g/mol): 250.28
Molecular Formula: C13H16NO4
InChi Key: LNKHBRDWRIIROP-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)