missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butoxycarbonylamino)cyclohexanone 98.0+%, TCI America™
$61.51 - $184.54
Chemical Identifiers
| CAS | 179321-49-4 |
|---|---|
| Molecular Formula | C11H19NO3 |
| Molecular Weight (g/mol) | 213.28 |
| MDL Number | MFCD00798168 |
| InChI Key | WYVFPGFWUKBXPZ-UHFFFAOYSA-N |
| Synonym | 4-(Boc-amino)cyclohexanone |
| PubChem CID | 1512535 |
| IUPAC Name | tert-butyl N-(4-oxocyclohexyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCC(=O)CC1 |
Chemical Identifiers
| 179321-49-4 | |
| 213.28 | |
| WYVFPGFWUKBXPZ-UHFFFAOYSA-N | |
| 1512535 | |
| CC(C)(C)OC(=O)NC1CCC(=O)CC1 |
| C11H19NO3 | |
| MFCD00798168 | |
| 4-(Boc-amino)cyclohexanone | |
| tert-butyl N-(4-oxocyclohexyl)carbamate |
Specifications
| 116°C | |
| White-Yellow | |
| MFCD00798168 | |
| WYVFPGFWUKBXPZ-UHFFFAOYSA-N | |
| tert-butyl N-(4-oxocyclohexyl)carbamate | |
| 213.28 | |
| 213.28 | |
| Crystalline Powder |
| 179321-49-4 | |
| C11H19NO3 | |
| 4-(Boc-amino)cyclohexanone | |
| CC(C)(C)OC(=O)NC1CCC(=O)CC1 | |
| 1 g | |
| 1512535 | |
| ≥98.0% (GC) | |
| 4-(tert-Butoxycarbonylamino)cyclohexanone |
Safety and Handling
TSCA : No