accessibility menu, dialog, popup

UserName

CAS RN 179321-49-4

H3CH3CCH3OONHO

CAS RN 179321-49-4

IUPAC Name: tert-butyl N-(4-oxocyclohexyl)carbamate
Synonyms: tert-butyl N-(4-oxocyclohexyl)carbamate
Molecular Weight (g/mol): 213.28
Molecular Formula: C11H19NO3
InChi Key: WYVFPGFWUKBXPZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(=O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results