missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butoxycarbonylamino)cyclohexanol 98.0+%, TCI America™
$61.51 - $188.27
Chemical Identifiers
| CAS | 224309-64-2 |
|---|---|
| Molecular Formula | C11H21NO3 |
| Molecular Weight (g/mol) | 215.29 |
| MDL Number | MFCD03844613,MFCD03844614,MFCD06658349 |
| InChI Key | DQARDWKWPIRJEH-UHFFFAOYSA-N |
| Synonym | 4-(Boc-amino)cyclohexanol, tert-Butyl (4-Hydroxycyclohexyl)carbamate, (4-Hydroxycyclohexyl)carbamic Acid tert-Butyl Ester |
| PubChem CID | 1514287 |
| IUPAC Name | tert-butyl N-(4-hydroxycyclohexyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCC(O)CC1 |
Chemical Identifiers
| 224309-64-2 | |
| 215.29 | |
| DQARDWKWPIRJEH-UHFFFAOYSA-N | |
| 1514287 | |
| CC(C)(C)OC(=O)NC1CCC(O)CC1 |
| C11H21NO3 | |
| MFCD03844613,MFCD03844614,MFCD06658349 | |
| 4-(Boc-amino)cyclohexanol, tert-Butyl (4-Hydroxycyclohexyl)carbamate, (4-Hydroxycyclohexyl)carbamic Acid tert-Butyl Ester | |
| tert-butyl N-(4-hydroxycyclohexyl)carbamate |
Specifications
| 224309-64-2 | |
| White-Yellow | |
| MFCD03844613,MFCD03844614,MFCD06658349 | |
| 4-(Boc-amino)cyclohexanol, tert-Butyl (4-Hydroxycyclohexyl)carbamate, (4-Hydroxycyclohexyl)carbamic Acid tert-Butyl Ester | |
| CC(C)(C)OC(=O)NC1CCC(O)CC1 | |
| 215.29 | |
| 215.29 | |
| Crystalline Powder |
| 172°C | |
| C11H21NO3 | |
| 1 g | |
| DQARDWKWPIRJEH-UHFFFAOYSA-N | |
| tert-butyl N-(4-hydroxycyclohexyl)carbamate | |
| 1514287 | |
| ≥98.0% (GC,N) | |
| 4-(tert-Butoxycarbonylamino)cyclohexanol |
Safety and Handling
TSCA : No