CAS RN 224309-64-2
IUPAC Name:
tert-butyl N-(4-hydroxycyclohexyl)carbamate
Synonyms:
tert-butyl N-(4-hydroxycyclohexyl)carbamate
Molecular Weight (g/mol):
215.29
Molecular Formula:
C11H21NO3
InChi Key:
DQARDWKWPIRJEH-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1CCC(O)CC1
1
–
1
of
1
results
1
4-(tert-Butoxycarbonylamino)cyclohexanol 98.0+%, TCI America™
CAS: 224309-64-2 Molecular Formula: C11H21NO3 Molecular Weight (g/mol): 215.29 MDL Number: MFCD03844613,MFCD03844614,MFCD06658349 InChI Key: DQARDWKWPIRJEH-UHFFFAOYSA-N Synonym: 4-(Boc-amino)cyclohexanol, tert-Butyl (4-Hydroxycyclohexyl)carbamate, (4-Hydroxycyclohexyl)carbamic Acid tert-Butyl Ester PubChem CID: 1514287 IUPAC Name: tert-butyl N-(4-hydroxycyclohexyl)carbamate SMILES: CC(C)(C)OC(=O)NC1CCC(O)CC1