missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonylamino)cyclohexanol 98.0+%, TCI America™

Catalog No. B47145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47145G 5 g
B47141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47145G Supplier TCI America Supplier No. B47145G
Only null left

Chemical Identifiers

CAS 224309-64-2
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD03844613,MFCD03844614,MFCD06658349
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym 4-(Boc-amino)cyclohexanol, tert-Butyl (4-Hydroxycyclohexyl)carbamate, (4-Hydroxycyclohexyl)carbamic Acid tert-Butyl Ester
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(O)CC1

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.29
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)cyclohexanol