missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$57.79 - $167.76
Chemical Identifiers
| CAS | 850568-54-6 |
|---|---|
| Molecular Formula | C11H15BO4 |
| MDL Number | MFCD03411946 |
| InChI Key | QMVMDYSTJSUDKC-UHFFFAOYSA-N |
| PubChem CID | 2773301 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4772200MG
|
TCI America
B4772200MG |
200 mg |
Each for $57.79
|
|
|||||
|
B47721G
|
TCI America
B47721G |
1 g |
Each for $167.76
|
|
|||||
Specifications
| 850568-54-6 | |
| White | |
| MFCD03411946 | |
| 4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid | |
| B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O | |
| 222.047 | |
| 222.05 | |
| 4-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
| 106°C | |
| C11H15BO4 | |
| 200 mg | |
| QMVMDYSTJSUDKC-UHFFFAOYSA-N | |
| [4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid | |
| 2773301 | |
| Crystalline Powder |
Safety and Handling
TSCA : No