accessibility menu, dialog, popup

UserName

CAS RN 850568-54-6

CH3H3CCH3OOBOHHO

CAS RN 850568-54-6

IUPAC Name: {4-[(tert-butoxy)carbonyl]phenyl}boronic acid
Synonyms: 4-(tert-butoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C11H15BO4
InChi Key: QMVMDYSTJSUDKC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results