missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfocalix[8]arene Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 1489.45 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Sulfocalix[8]arene Hydrate |
Safety and Handling
| TSCA | No |