missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfamoylbenzoic Acid 95.0+%, TCI America™

Catalog No. p-7135861
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S032325G 25 g
S0323500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S032325G Supplier TCI America Supplier No. S032325G
Only null left

Chemical Identifiers

CAS 138-41-0
Molecular Formula C7H7NO4S
Molecular Weight (g/mol) 201.196
MDL Number MFCD00007938
InChI Key UCAGLBKTLXCODC-UHFFFAOYSA-N
Synonym carzenide, 4-carboxybenzenesulfonamide, 4-aminosulfonyl benzoic acid, carzenid, dirnate, p-carboxybenzenesulfonamide, benzoic acid, 4-aminosulfonyl, 4-aminosulfonylbenzoic acid, p-sulfamylbenzoic acid, p-sulfamoylbenzoic acid
PubChem CID 8739
IUPAC Name 4-sulfamoylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 201.20
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfamoylbenzoic Acid
EINECSNumber (3)-2109
RTECSNumber DH6820000
TSCA Yes