missing translation for 'onlineSavingsMsg'
Learn More

4-Styrylpyridine 98.0+%, TCI America™

$125.13 - $375.41

Chemical Identifiers

CAS 103-31-1
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.24
MDL Number MFCD00137749
InChI Key QKHRGPYNTXRMSL-VOTSOKGWSA-N
Synonym 4-Stilbazole
PubChem CID 641098
IUPAC Name 4-[(1E)-2-phenylethenyl]pyridine
SMILES C(=C/C1=CC=NC=C1)\C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Qty  
S04751G
View Documents
TCI America
S04751G
1 g
Each for $125.13
Only null left
 
S04755G
View Documents
TCI America
S04755G
5 g
Each for $375.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

103-31-1
181.24
QKHRGPYNTXRMSL-VOTSOKGWSA-N
641098
C(=C/C1=CC=NC=C1)\C1=CC=CC=C1
C13H11N
MFCD00137749
4-Stilbazole
4-[(1E)-2-phenylethenyl]pyridine

Specifications

103-31-1
C13H11N
1 g
QKHRGPYNTXRMSL-VOTSOKGWSA-N
4-[(1E)-2-phenylethenyl]pyridine
641098
≥98.0% (GC,T)
4-Styrylpyridine
131°C
MFCD00137749
4-Stilbazole
C(=C/C1=CC=NC=C1)\C1=CC=CC=C1
181.24
181.24
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents